Learn More
Chemglass Life Sciences Condenser, Allihn, 335mm, 29/42 Joint, 200mm Jacket Length
SDP
Catalog No. 50873018
Encompass_Preferred
Condenser, Allihn, 335mm, 29/42 Joint, 200mm Jacket Length. Allihn style reflux condenser with a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. 50-873-018 Supplier Chemglass Life Sciences Supplier No. CG120602
Condenser, Allihn, 335mm, 29/42 Joint, 200mm Jacket Length. Allihn style reflux condenser with a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.