Learn More
Chemglass Life Sciences Condenser, Allihn, 420mm OAH, Static Waterless, 24/40 Jt, 300mm Jacket Length
SDP
Catalog No. NC1718607
Encompass_Preferred
Condenser, Allihn, 420mm OAH, Static Waterless, 24/40 Jt, 300mm
Catalog No. NC1718607 Supplier Chemglass Life Sciences Supplier No. CG1206W224
Condenser, Allihn, 420mm OAH, Static Waterless, 24/40 Jt, 300mm