Learn More
Ace Glass, Inc. Condenser, bullfrog, poly-coated, model R220, glassware sets DB&DB2, part 27825
SDP
Catalog No. 50971488
Encompass
Borosilicate glass replacement poly-coated bullfrog condenser for use in rotary evaporator Model R220 glassware sets DB & DB2; part 27825. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-971-488 Supplier Ace Glass, Inc. Supplier No. 396519
Add to Cart
Add to Cart
Request a Quote
Borosilicate glass replacement poly-coated bullfrog condenser for use in rotary evaporator Model R220 glassware sets DB & DB2; part 27825. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.