Learn More
Ace Glass, Inc. Condenser, bullfrog, poly-coated, model R220, glassware setsRB&DB2, part 27824
SDP
Catalog No. 50978236
Encompass
Borosilicate glass replacement poly-coated bullfrog condenser for use in rotary evaporator Model R220 glassware sets RB & DB2; part 27824. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-978-236 Supplier Ace Glass, Inc. Supplier No. 396526
Add to Cart
Add to Cart
Request a Quote
Borosilicate glass replacement poly-coated bullfrog condenser for use in rotary evaporator Model R220 glassware sets RB & DB2; part 27824. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.