Learn More
Ace Glass, Inc. Condenser, bullfrog, models R220EX&SE, glassware sets DB&DB2, part 46516
SDP
Catalog No. 50971489
Encompass
Borosilicate glass replacement bullfrog condenser for use in rotary evaporator Model R220EX&SE glassware sets DB & DB2; part 46516. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-971-489 Supplier Ace Glass, Inc. Supplier No. 396522
Add to Cart
Add to Cart
Request a Quote
Borosilicate glass replacement bullfrog condenser for use in rotary evaporator Model R220EX&SE glassware sets DB & DB2; part 46516. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.