Learn More
Ace Glass, Inc. Condenser, bullfrog, model R220, glassware sets RB&DB2, part 41458
SDP
Catalog No. 50971490
Encompass
Borosilicate glass replacement bullfrog condenser for use in rotary evaporator Model R220 glassware sets RB & DB2; part 41458. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.
Catalog No. 50-971-490 Supplier Ace Glass, Inc. Supplier No. 396528
Add to Cart
Add to Cart
Request a Quote
Borosilicate glass replacement bullfrog condenser for use in rotary evaporator Model R220 glassware sets RB & DB2; part 41458. Inner ball joints include CAPFE o-rings. Includes black SVL caps & red GL thread caps.