Learn More
Chemglass Life Sciences Condenser, "C" Assembly, For Rotary Evaporator, Plastic Coated
SDP
Catalog No. CG133310
Encompass_Preferred
Condenser, "C" Assembly, For Rotary Evaporator, Plastic Coated. Two piece cold trap style condenser for use with rotary evaporators. Condenser has a 50mm flange on the side, a 35/20 lower ball joint, a 19/38 stopcock plug and removable hose connections using GL-14 threads. Condenser is supplied complete with PTFE centering ring along with plastic caps and serrated hose connections. Plastic coating is on outer portion of the condenser only.
Catalog No. CG133310 Supplier Chemglass Life Sciences Supplier No. CG133310
Condenser, "C" Assembly, For Rotary Evaporator, Plastic Coated. Two piece cold trap style condenser for use with rotary evaporators. Condenser has a 50mm flange on the side, a 35/20 lower ball joint, a 19/38 stopcock plug and removable hose connections using GL-14 threads. Condenser is supplied complete with PTFE centering ring along with plastic caps and serrated hose connections. Plastic coating is on outer portion of the condenser only.