Learn More
Chemglass Life Sciences Condenser, "C" Assembly, Constant Feed, For Rotary Evaporators, Plastic Coated
SDP
Catalog No. CG1333A01
Encompass_Preferred
Condenser, "C" Assembly, Constant Feed, For Rotary Evaporators, Plastic Coated. Two piece cold trap style condenser for use with rotary evaporators. Condenser has an additional 19/38 outer joint sealed onto the condenser to allow continuous feed of volumes exceeding the capacity of the evaporating flask. The 19/38 joint is sealed at an angle. This continuous feed feature eliminates the need for a separate filtration step to remove drying agents or other particulate matter from organic solutions prior to rotary evaporation. The ferrule style connection eliminates o-ring connections, which are prone to degradation with long term exposure to organic solvents. Condenser is supplied complete with all fittings necessary to connect the continuous feed feature, along with our new style PTFE centering ring which makes a vacuum tight seal between the inner cold finger and the plastic coated outer sleeve. Also supplied with a UHMW lid with knob.
Catalog No. CG1333A01 Supplier Chemglass Life Sciences Supplier No. CG1333A01
Condenser, "C" Assembly, Constant Feed, For Rotary Evaporators, Plastic Coated. Two piece cold trap style condenser for use with rotary evaporators. Condenser has an additional 19/38 outer joint sealed onto the condenser to allow continuous feed of volumes exceeding the capacity of the evaporating flask. The 19/38 joint is sealed at an angle. This continuous feed feature eliminates the need for a separate filtration step to remove drying agents or other particulate matter from organic solutions prior to rotary evaporation. The ferrule style connection eliminates o-ring connections, which are prone to degradation with long term exposure to organic solvents. Condenser is supplied complete with all fittings necessary to connect the continuous feed feature, along with our new style PTFE centering ring which makes a vacuum tight seal between the inner cold finger and the plastic coated outer sleeve. Also supplied with a UHMW lid with knob.