Learn More
Ace Glass, Inc. Condenser, coiled, 24/40 top & bottom, 200mm jacket, Ace-Safe hose connections for 1/4in ID tubing
SDP
Catalog No. NC1943072
Encompass_Preferred
Coiled condensing tube, with 24/40 inner and outer joint at bottom and top. 200mm jacket,Ace-Safe hose connections for 1/4in ID tubing.
Catalog No. NC1943072 Supplier Ace Glass, Inc. Supplier No. 5953101
Coiled condensing tube, with 24/40 inner and outer joint at bottom and top. 200mm jacket,Ace-Safe hose connections for 1/4in ID tubing.