Learn More
Ace Glass, Inc. Condenser, coiled, reflux, 300mm, 24/40 st joints top and bottom
SDP
Catalog No. 50972669
Encompass_Preferred
300mm double-jacketed, coiled reflux condenser with 24/40 standard inner and outer joints at bottom and top.
Catalog No. 50-972-669 Supplier Ace Glass, Inc. Supplier No. 595514
300mm double-jacketed, coiled reflux condenser with 24/40 standard inner and outer joints at bottom and top.