Learn More
Ace Glass, Inc. Condenser, coiled, reflux, 400mm, 24/40 st joints top and bottom
SDP
Catalog No. 50972670
Encompass_Preferred
400mm double-jacketed, coiled reflux condenser with 24/40 standard inner and outer joints at bottom and top.
Catalog No. 50-972-670 Supplier Ace Glass, Inc. Supplier No. 595515
Add to Cart
Add to Cart
400mm double-jacketed, coiled reflux condenser with 24/40 standard inner and outer joints at bottom and top.