Learn More
Chemglass Life Sciences Condenser, Cold Finger, Adjustable, Minum-Ware
SDP
Catalog No. 501532679
Encompass_Preferred
Condenser, Cold Finger, Adjustable, Minum-Ware. Cold finger style condenser is made to fit into MW-50 Hickman still head while the lower drip tip directs distillate into the collection collar. Complete item consists of the condenser and a MW-33 inlet adapter having a 14/10 lower inner joint. Hose connections have an O.D. of 9mm at the middle serration.
Catalog No. 50-153-2679 Supplier Chemglass Life Sciences Supplier No. MW4301
Condenser, Cold Finger, Adjustable, Minum-Ware. Cold finger style condenser is made to fit into MW-50 Hickman still head while the lower drip tip directs distillate into the collection collar. Complete item consists of the condenser and a MW-33 inlet adapter having a 14/10 lower inner joint. Hose connections have an O.D. of 9mm at the middle serration.