Learn More
Chemglass Life Sciences Condenser, Cold Finger, 29/42 Inner Joint, 200mm Length Below Joint
SDP
Catalog No. CG122204
Encompass_Preferred
Condenser, Cold Finger, 29/42 Inner Joint, 200mm Length Below Joint. Condenser has an inner joint and a sealed lower drip tip for drop control. Hose connections have an O.D. of 10mm at the middle serration
Catalog No. CG122204 Supplier Chemglass Life Sciences Supplier No. CG122204
Condenser, Cold Finger, 29/42 Inner Joint, 200mm Length Below Joint. Condenser has an inner joint and a sealed lower drip tip for drop control. Hose connections have an O.D. of 10mm at the middle serration