Learn More
Chemglass Life Sciences Condenser, Concentrator, 325mm, 24/40 Joint, 200mm Inner Tube Height
SDP
Catalog No. CG120820
Encompass_Preferred
Condenser, Concentrator, 325mm, 24/40 Joint, 200mm Inner Tube Height. Vapors rise to the top of inner tube then travel downward and condense in this highly efficient concentrator. Condensate drains through the side tube which has a standard taper inner joint the same size as the lower inner uptake joint. Serrated hose connection for connecting to vacuum source. Hose connections for both the cooling jacket and vacuum source have an O.D. of 10mm. Available with detachable hose connections on a special order basis.
Catalog No. CG120820 Supplier Chemglass Life Sciences Supplier No. CG120820
Condenser, Concentrator, 325mm, 24/40 Joint, 200mm Inner Tube Height. Vapors rise to the top of inner tube then travel downward and condense in this highly efficient concentrator. Condensate drains through the side tube which has a standard taper inner joint the same size as the lower inner uptake joint. Serrated hose connection for connecting to vacuum source. Hose connections for both the cooling jacket and vacuum source have an O.D. of 10mm. Available with detachable hose connections on a special order basis.