Learn More
Chemglass Life Sciences Condenser, ''CR'' Assembly, For Rotary Evaporators, Plastic Coated
SDP
Catalog No. CG133510
Encompass_Preferred
Condenser, ''CR'' Assembly, For Rotary Evaporators, Plastic Coated. Two piece cold trap style condenser for use with rotary evaporators. Condenser has a 45/40 lower inner drip joint, a 19/38 stopcock plug, and removable hose connections using GL-14 threads. Condenser is supplied complete with PTFE centering ring along with plastic caps and serrated hose connections. Plastic coating is on the outer portion of condenser only.
Catalog No. CG133510 Supplier Chemglass Life Sciences Supplier No. CG133510
Condenser, ''CR'' Assembly, For Rotary Evaporators, Plastic Coated. Two piece cold trap style condenser for use with rotary evaporators. Condenser has a 45/40 lower inner drip joint, a 19/38 stopcock plug, and removable hose connections using GL-14 threads. Condenser is supplied complete with PTFE centering ring along with plastic caps and serrated hose connections. Plastic coating is on the outer portion of condenser only.