Learn More
Chemglass Life Sciences Condenser, Dewar Type, 24/40 Inner Joint, 200 x 40mm
SDP
Catalog No. CG120901
Encompass_Preferred
Condenser, Dewar Type, 24/40 Inner Joint, 200 x 40mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has a lower inner drip tip joint and hose connections with an O.D. of 10mm at the middle serration.• NOTE: See CG-4519 For Solvent Recovery Traps used with Rotary Evaporators.
Catalog No. CG120901 Supplier Chemglass Life Sciences Supplier No. CG120901
Condenser, Dewar Type, 24/40 Inner Joint, 200 x 40mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has a lower inner drip tip joint and hose connections with an O.D. of 10mm at the middle serration.• NOTE: See CG-4519 For Solvent Recovery Traps used with Rotary Evaporators.