Learn More
Ace Glass, Inc. Condenser, double coil, 1in beaded pipe top and bottom joints
SDP
Supplier:  Ace Glass, Inc. 601512
Encompass
Double coil condenser with 1 beaded pipe top and bottom joints. Supplied with #15 Ace-Thred connections for Ace-Safe hose barbs. Unit length is approximately 390mm and the O.D. is approximately 85-90mm. Condenser is designed to fit larger systems and pilot plant reactors.
Catalog No. 50-972-709