Learn More
Ace Glass, Inc. Condenser, double coil, 45/50 outer top, 45/50 inner bottom
SDP
Catalog No. 50972710
Encompass
Double coil condenser with 45/50 outer top and 45/50 inner bottom joints. Supplied with #15 Ace-Thred connections for Ace-Safe hose barbs. Unit length is approximately 390mm and the O.D. is approximately 85-90mm. Condenser is designed to fit larger systems and pilot plant reactors.
Catalog No. 50-972-710 Supplier Ace Glass, Inc. Supplier No. 601517
Double coil condenser with 45/50 outer top and 45/50 inner bottom joints. Supplied with #15 Ace-Thred connections for Ace-Safe hose barbs. Unit length is approximately 390mm and the O.D. is approximately 85-90mm. Condenser is designed to fit larger systems and pilot plant reactors.