Learn More
Chemglass Life Sciences Condenser, Double Coil, 895cm², High Efficiency, 45/50 Joints, 400 x 80mm, GL-14 Screw Caps, Removable Hose Connections
SDP
Catalog No. CG121920
Encompass_Preferred
Condenser, Double Coil, 895cm¿, High Efficiency, 45/50 Joints, 400 x 80mm, GL-14 Screw Caps, Removable Hose Connections. High efficiency double coil condenser, similar to a rotary evaporator condenser but with 45/50 joints. Condenser has an O.D. of 80mm and is 400mm overall length. Coolant connections are made using removable hose connections with GL-14 threads. Supplied complete with two polypropylene hose connections and caps.
Catalog No. CG121920 Supplier Chemglass Life Sciences Supplier No. CG121920
Condenser, Double Coil, 895cm¿, High Efficiency, 45/50 Joints, 400 x 80mm, GL-14 Screw Caps, Removable Hose Connections. High efficiency double coil condenser, similar to a rotary evaporator condenser but with 45/50 joints. Condenser has an O.D. of 80mm and is 400mm overall length. Coolant connections are made using removable hose connections with GL-14 threads. Supplied complete with two polypropylene hose connections and caps.