Learn More
Chemglass Life Sciences Condenser, Friedrichs, 335mm, 29/42 Joint
SDP
Catalog No. CG121002
Encompass_Preferred
Condenser, Friedrichs, 335mm, 29/42 Joint. Condenser has a specially formed spiral inner tube providing a long vapor path, ensuring good heat transfer and anti-flooding characteristics. Condenser has a standard taper outer joint at a 15º angle to the condenser and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG121002 Supplier Chemglass Life Sciences Supplier No. CG121002
Condenser, Friedrichs, 335mm, 29/42 Joint. Condenser has a specially formed spiral inner tube providing a long vapor path, ensuring good heat transfer and anti-flooding characteristics. Condenser has a standard taper outer joint at a 15º angle to the condenser and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.