Learn More
Chemglass Life Sciences Condenser, Friedrichs, 360mm, 45/50 Inner Joint
SDP
Catalog No. CG121103
Encompass_Preferred
Condenser, Friedrichs, 360mm, 45/50 Inner Joint. Similar to CG-1210 but with the 24/40 outer joint replaced with a serrated hose connection for use with a drying tube. This style condenser is used primarily in Soxhlet extractions or in reflux mode. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG121103 Supplier Chemglass Life Sciences Supplier No. CG121103
Condenser, Friedrichs, 360mm, 45/50 Inner Joint. Similar to CG-1210 but with the 24/40 outer joint replaced with a serrated hose connection for use with a drying tube. This style condenser is used primarily in Soxhlet extractions or in reflux mode. Hose connections have an O.D. of 10mm at the middle serration.