Learn More
Chemglass Life Sciences Condenser, Reflux, 24/40, 345mm OAH
SDP
Catalog No. 50143905
Encompass_Preferred
Coil stlye reflux condenser with a larger diameter jacket and coil, thereby providing maximum cooling capacity, while being shorter in length. Hose connections are on the bottom of the condenser which makes it easier to connect hose/tubing. Connections are GL14 polypropylene hose barbs and screw caps. GL hose connections provide a safe and easy way of connecting flexible tubing to a condenser. Jacket has a O. D. of 41mm while the coil O. D. is 8mm. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Catalog No. 50-143-905 Supplier Chemglass Life Sciences Supplier No. CG1213JHC01
Coil stlye reflux condenser with a larger diameter jacket and coil, thereby providing maximum cooling capacity, while being shorter in length. Hose connections are on the bottom of the condenser which makes it easier to connect hose/tubing. Connections are GL14 polypropylene hose barbs and screw caps. GL hose connections provide a safe and easy way of connecting flexible tubing to a condenser. Jacket has a O. D. of 41mm while the coil O. D. is 8mm. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.