Learn More
Chemglass Life Sciences Chemglass Bulb-Style Glass Condenser, 150 mm OAL
SDP
Catalog No. 502733319
Encompass
Glass bulb-style condenser designed to remove moisture from exhaust gases and minimize evaporation loss in laboratory bioreactors. Compact 150 mm overall length makes it suitable for small-scale fermentation and bioprocessing setups.
  • Provides efficient condensation to minimize evaporation losses.
  • 150 mm overall length for compact installations.
  • Bulb-style design increases condensation surface area.
  • Constructed from glass for chemical resistance and visibility.
  • Pairs with standard bioreactor adapters or probe adapters (sold separately).
Catalog No. 50-273-3319 Supplier Chemglass Life Sciences Supplier No. CLS1380GC1
Glass bulb-style condenser designed to remove moisture from exhaust gases and minimize evaporation loss in laboratory bioreactors. Compact 150 mm overall length makes it suitable for small-scale fermentation and bioprocessing setups.
  • Provides efficient condensation to minimize evaporation losses.
  • 150 mm overall length for compact installations.
  • Bulb-style design increases condensation surface area.
  • Constructed from glass for chemical resistance and visibility.
  • Pairs with standard bioreactor adapters or probe adapters (sold separately).