Learn More
Chemglass Life Sciences Condenser, Glass, 250mm OAL, Bulb-Style, for 7L Bioreactors
SDP
Catalog No. 501294442
Encompass
Condenser, Glass, 250mm OAL, Bulb-Style, for 7L Bioreactors. Condensers aid in removing moisture from exhaust gases which can clog filters. They also help minimize loss of material from the bioreactor due to evaporation during fermentation. Glass condensers are available in bulb and coil styles and are more economical than stainless steel. Stainless steel condensers consist of 4 components: 1. Jacketed condenser body with two 1/4-inch hose connections 2. Internal stainless steel baffle 3. 90o gas outlet tube with 1/4-inch hose connection 3. Threaded compression nut All condensers are connected to the bioreactor by using 12mm adjustable height probe adapters which are sold separately.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4442 Supplier Chemglass Life Sciences Supplier No. CLS1380GC2
Condenser, Glass, 250mm OAL, Bulb-Style, for 7L Bioreactors. Condensers aid in removing moisture from exhaust gases which can clog filters. They also help minimize loss of material from the bioreactor due to evaporation during fermentation. Glass condensers are available in bulb and coil styles and are more economical than stainless steel. Stainless steel condensers consist of 4 components: 1. Jacketed condenser body with two 1/4-inch hose connections 2. Internal stainless steel baffle 3. 90o gas outlet tube with 1/4-inch hose connection 3. Threaded compression nut All condensers are connected to the bioreactor by using 12mm adjustable height probe adapters which are sold separately.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.