Learn More
Ace Glass, Inc. Condenser, Graham, 900mm, 24/40
SDP
Catalog No. 50978637
Encompass_Preferred
900mm Graham condenser with 24/40 standard taper inner and outer joint at top and bottom. Hose connections are size D for 3/8 ID tubing.
Catalog No. 50-978-637 Supplier Ace Glass, Inc. Supplier No. 597719
900mm Graham condenser with 24/40 standard taper inner and outer joint at top and bottom. Hose connections are size D for 3/8 ID tubing.