Learn More
Chemglass Life Sciences Chemglass high-efficiency double cooling coil distillation condenser, 65/40 top socket, angled 45/50 inner joint (CG-1968-D-306)
SDP
Catalog No. 502733551
Encompass_Preferred
High-efficiency double cooling coil condenser designed for distillation and reflux in process reactor and distillation cart systems. It incorporates a 65/40 socket at the top, an angled 45/50 inner joint at the bottom, hose connections on the inlet/outlet, and a vacuum take-off for reduced-pressure operation.
  • Double cooling coil design improves condensate return and heat exchange.
  • 65/40 top socket and angled 45/50 bottom joint for compatibility with standard reactor fittings.
  • Hose connections on inlet and outlet for coolant circulation.
  • Vacuum take-off supports reduced-pressure distillation.
  • Compact configuration suitable for integration into distillation carts and reactor assemblies.
Catalog No. 50-273-3551 Supplier Chemglass Life Sciences Supplier No. CG1968D306
High-efficiency double cooling coil condenser designed for distillation and reflux in process reactor and distillation cart systems. It incorporates a 65/40 socket at the top, an angled 45/50 inner joint at the bottom, hose connections on the inlet/outlet, and a vacuum take-off for reduced-pressure operation.
  • Double cooling coil design improves condensate return and heat exchange.
  • 65/40 top socket and angled 45/50 bottom joint for compatibility with standard reactor fittings.
  • Hose connections on inlet and outlet for coolant circulation.
  • Vacuum take-off supports reduced-pressure distillation.
  • Compact configuration suitable for integration into distillation carts and reactor assemblies.