Learn More
Chemglass Life Sciences Condenser, Jacketed Reflux, Minum-Ware
SDP
Catalog No. 50873031
Encompass_Preferred
Condenser, Jacketed Reflux, Minum-Ware. Liebig-type water jacketed condenser with a jacket length of 80mm. With top threaded outer and lower inner drip joint, both 14/10. Supplied complete with open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).
Catalog No. 50-873-031 Supplier Chemglass Life Sciences Supplier No. MW4201
Condenser, Jacketed Reflux, Minum-Ware. Liebig-type water jacketed condenser with a jacket length of 80mm. With top threaded outer and lower inner drip joint, both 14/10. Supplied complete with open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).