Learn More
Heidolph™ Condenser for Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware
SDP
Catalog No. 13900919 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota 20, Hei-VAP Industrial Rotary Evaporator
Catalog No. For Use With (Equipment)
13-900-919 Laborota 20, Hei-VAP Industrial Rotary Evaporator
1 options
Catalog No. 13-900-919 Supplier Heidolph™ Supplier No. 036304421
Add to Cart
Add to Cart
Condenser allows distillate to re-condense. Recommended for all standard distillations
Included with Laborota 20, Hei-VAP Industrial Rotary Evaporator Glassware R and shorty R
Specifications
Type Glassware
For Use With (Equipment) Laborota 20, Hei-VAP Industrial Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov