Learn More
Chemglass Life Sciences Condenser, Liebig, 325mm, 29/42 Joint, 200mm Jacket Length
SDP
Catalog No. CG121802
Encompass_Preferred
Joint, 28/12, Polished Socket, Clamp Size 28, 16mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.
Catalog No. CG121802 Supplier Chemglass Life Sciences Supplier No. CG121802
Joint, 28/12, Polished Socket, Clamp Size 28, 16mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.