Learn More
Chemglass Life Sciences Condenser, Liebig, 525mm, 29/42 Joint, 400mm Jacket Length
SDP
Catalog No. CG121808
Encompass_Preferred
Condenser, Liebig, 525mm, 29/42 Joint, 400mm Jacket Length. Condenser has a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.
Catalog No. CG121808 Supplier Chemglass Life Sciences Supplier No. CG121808
Condenser, Liebig, 525mm, 29/42 Joint, 400mm Jacket Length. Condenser has a standard taper outer joint at the top and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration.