Learn More
Ace Glass, Inc. Condenser, Liebig, 200mm, 19/38
SDP
Catalog No. 50972699
Encompass_Preferred
200mm Liebig condenser with 19/38 standard taper or spherical joint at bottom and top. Hose connections are size D.
Catalog No. 50-972-699 Supplier Ace Glass, Inc. Supplier No. 599804
200mm Liebig condenser with 19/38 standard taper or spherical joint at bottom and top. Hose connections are size D.