Learn More
Ace Glass, Inc. Condenser, Liebig, 200mm, 29/42
SDP
Catalog No. 50972703
Encompass_Preferred
200mm Liebig condenser with 29/42 standard taper or spherical joint at bottom and top. Hose connections are size D.
Catalog No. 50-972-703 Supplier Ace Glass, Inc. Supplier No. 599822
200mm Liebig condenser with 29/42 standard taper or spherical joint at bottom and top. Hose connections are size D.