Learn More
Ace Glass, Inc. Condenser, Liebig, no hold up, 200mm, 24/40 joints top & bottom, inlet/outlet #7 Ace-Thred connections, complete with Ace-Safe connectors
SDP
Catalog No. NC1943069
Encompass_Preferred
Condenser, Liebig, no hold up, 200mm, 24/40 joints top & bottom, inlet/outlet #7 Ace-Thred connections, complete with Ace-Safe connectors
Catalog No. NC1943069 Supplier Ace Glass, Inc. Supplier No. 5997132
Condenser, Liebig, no hold up, 200mm, 24/40 joints top & bottom, inlet/outlet #7 Ace-Thred connections, complete with Ace-Safe connectors