Learn More
Ace Glass, Inc. Condenser, Liebig, 400mm, 29/42
SDP
Catalog No. 50972705
Encompass_Preferred
400mm Liebig condenser with 29/42 standard taper or spherical joint at bottom and top. Hose connections are size D.
Catalog No. 50-972-705 Supplier Ace Glass, Inc. Supplier No. 599825
400mm Liebig condenser with 29/42 standard taper or spherical joint at bottom and top. Hose connections are size D.