Learn More
Ace Glass, Inc. Condenser, Liebig, 500mm, 24/40
SDP
Catalog No. 50978640
Encompass_Preferred
500mm Liebig condenser with 24/40 standard taper or spherical joint at bottom and top. Hose connections are size D.
Catalog No. 50-978-640 Supplier Ace Glass, Inc. Supplier No. 599816
500mm Liebig condenser with 24/40 standard taper or spherical joint at bottom and top. Hose connections are size D.