Learn More
Ace Glass, Inc. Condenser, Liebig, 600mm, 24/40
SDP
Catalog No. 50972702
Encompass_Preferred
600mm Liebig condenser with 24/40 standard taper or spherical joint at bottom and top. Hose connections are size D.
Catalog No. 50-972-702 Supplier Ace Glass, Inc. Supplier No. 599817
600mm Liebig condenser with 24/40 standard taper or spherical joint at bottom and top. Hose connections are size D.