Learn More
Ace Glass, Inc. Condenser, 250mm, 24/40 st joints top and bottom, #7 Ace-Thred connections, glassware only
SDP
Catalog No. 50978630
Encompass_Preferred
250mm double-jacketed, coiled condenser with standard 24/40 inner and outer joint at bottom and top. Includes #7 Ace-Thred connections. Provides extra cooling capacity for volatile materials.
Catalog No. 50-978-630 Supplier Ace Glass, Inc. Supplier No. 595643
250mm double-jacketed, coiled condenser with standard 24/40 inner and outer joint at bottom and top. Includes #7 Ace-Thred connections. Provides extra cooling capacity for volatile materials.