Learn More
Chemglass Life Sciences Large Condenser Only, Extraction Apparatus, Soxhlet, 55/50
SDP
Catalog No. 50873029
Encompass_Preferred
Large condenser. Part of the CG1368 series. Condenser has an enlarged uptake tube to improve condensing capacity.
Catalog No. 50-873-029 Supplier Chemglass Life Sciences Supplier No. CG136806
Large condenser. Part of the CG1368 series. Condenser has an enlarged uptake tube to improve condensing capacity.