Learn More
Ace Glass, Inc. Condenser, Liebig, no hold up, 250mm, 24/40 joints top & bottom, inlet/outlet #7 Ace-Thred connections, glass only
SDP
Catalog No. 50972696
Encompass_Preferred
Condenser, Liebig, no hold up, 250mm, 24/40 joints top & bottom, inlet/outlet #7 Ace-Thred connections, glass only
Catalog No. 50-972-696 Supplier Ace Glass, Inc. Supplier No. 599733
Condenser, Liebig, no hold up, 250mm, 24/40 joints top & bottom, inlet/outlet #7 Ace-Thred connections, glass only