Learn More
Ace Glass, Inc. Condenser, Liebig, no hold up, 300mm, 24/40 joints top & bottom, inlet/outlet #7 Ace-Thred connections, glass only
SDP
Catalog No. 50972697
Encompass_Preferred
Condenser, Liebig, no hold up, 300mm, 24/40 joints top & bottom, inlet/outlet #7 Ace-Thred connections, glass only
Catalog No. 50-972-697 Supplier Ace Glass, Inc. Supplier No. 599734
Condenser, Liebig, no hold up, 300mm, 24/40 joints top & bottom, inlet/outlet #7 Ace-Thred connections, glass only