Learn More
Ace Glass, Inc. Condenser, spiral, reflux, 100mm, 14/38
SDP
Catalog No. 50972738
Encompass_Preferred
100mm 14/38 Spiral reflux condenser with inlet and outlet hose barbs located on the same side at the top of the condenser body for easy hose connections
Catalog No. 50-972-738 Supplier Ace Glass, Inc. Supplier No. 6041102
100mm 14/38 Spiral reflux condenser with inlet and outlet hose barbs located on the same side at the top of the condenser body for easy hose connections