Learn More
Chemglass Life Sciences Condenser, Reflux, 210mm, 14/20 Joint Size, 100mm Jacket Length, Removable Hose Connections
SDP
Catalog No. 50873023
Encompass_Preferred
Condenser, Reflux, 210mm, 14/20 Joint Size, 100mm Jacket Length, Removable Hose Connections. Reflux condenser similar to CG-1213-A but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Hose connections have an O.D. of 8mm and an I.D. of 4mm. Supplied complete with two GL-14 open top screw caps. See CG-198 and CG-195 for replacement GL fittings.
Catalog No. 50-873-023 Supplier Chemglass Life Sciences Supplier No. CG1213AHC01
Condenser, Reflux, 210mm, 14/20 Joint Size, 100mm Jacket Length, Removable Hose Connections. Reflux condenser similar to CG-1213-A but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Hose connections have an O.D. of 8mm and an I.D. of 4mm. Supplied complete with two GL-14 open top screw caps. See CG-198 and CG-195 for replacement GL fittings.