Learn More
Chemglass Life Sciences Condenser, Reflux, 350mm, 24/40 Joint, 175mm Coil Length, GL-14 Screw Cap, Removable Hose Connections
SDP
Catalog No. CG1213LHC02
Encompass_Preferred
Condenser, Reflux, 350mm, 24/40 Joint, 175mm Coil Length, GL-14 Screw Cap, Removable Hose Connections. Coil style reflux condenser with a large diameter jacket and coil, similar to CG-1213-L but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Supplied complete with two GL open top screw caps and two bent polypropylene hose connections. Condensing area is the same as CG-1213-L.
Catalog No. CG1213LHC02 Supplier Chemglass Life Sciences Supplier No. CG1213LHC02
Add to Cart
Add to Cart
Condenser, Reflux, 350mm, 24/40 Joint, 175mm Coil Length, GL-14 Screw Cap, Removable Hose Connections. Coil style reflux condenser with a large diameter jacket and coil, similar to CG-1213-L but with detachable hose connections. Connections are made of polypropylene and provide a safe and easy way of connecting and disconnecting flexible tubing to the condenser. Supplied complete with two GL open top screw caps and two bent polypropylene hose connections. Condensing area is the same as CG-1213-L.