Learn More
Chemglass Life Sciences Condenser, Reflux, 300mm, 24/40 Joint, 125mm Coil Length
SDP
Catalog No. CG1213L20
Encompass_Preferred
Condenser, Reflux, 300mm, 24/40 Joint, 125mm Coil Length. Similar to CG-1213, but having a larger diameter jacket and coil, thereby providing maximum cooling capacity, while being shorter in length. Jacket has an O.D. of 41mm while the coil O.D. is 8mm. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Catalog No. CG1213L20 Supplier Chemglass Life Sciences Supplier No. CG1213L20
Add to Cart
Add to Cart
Condenser, Reflux, 300mm, 24/40 Joint, 125mm Coil Length. Similar to CG-1213, but having a larger diameter jacket and coil, thereby providing maximum cooling capacity, while being shorter in length. Jacket has an O.D. of 41mm while the coil O.D. is 8mm. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.