Learn More
Chemglass Life Sciences Condenser, Reflux, 350mm, 24/40 Joint, 175mm Coil Length
SDP
Catalog No. CG1213L21
Encompass_Preferred
Condenser, Reflux, 350mm, 24/40 Joint, 175mm Coil Length. Similar to CG-1213, but having a larger diameter jacket and coil, thereby providing maximum cooling capacity, while being shorter in length. Jacket has an O.D. of 41mm while the coil O.D. is 8mm. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Catalog No. CG1213L21 Supplier Chemglass Life Sciences Supplier No. CG1213L21
Condenser, Reflux, 350mm, 24/40 Joint, 175mm Coil Length. Similar to CG-1213, but having a larger diameter jacket and coil, thereby providing maximum cooling capacity, while being shorter in length. Jacket has an O.D. of 41mm while the coil O.D. is 8mm. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.