Learn More
Ace Glass, Inc. Condenser, spiral, reflux, 300mm, 24/29
SDP
Catalog No. 50978653
Encompass_Preferred
300mm 24/29 Spiral reflux condenser with inlet and outlet hose barbs located on the same side at the top of the condenser body for easy hose connections
Catalog No. 50-978-653 Supplier Ace Glass, Inc. Supplier No. 6041108
300mm 24/29 Spiral reflux condenser with inlet and outlet hose barbs located on the same side at the top of the condenser body for easy hose connections