Learn More
Ace Glass, Inc. Condenser, reflux spiral, 300mm, 24/40 joints, size-d, 3/8-in hose connections
SDP
Catalog No. 50972726
Encompass_Preferred
300mm condenser with a reflux spiral condensing tube having both 3/8 inlet and outlet hose connections at top. Also features 24/40 standard taper joints. Length between joints is approx. 80-90mm longer than the stated coil length.
Catalog No. 50-972-726 Supplier Ace Glass, Inc. Supplier No. 602006
300mm condenser with a reflux spiral condensing tube having both 3/8 inlet and outlet hose connections at top. Also features 24/40 standard taper joints. Length between joints is approx. 80-90mm longer than the stated coil length.