Learn More
Ace Glass, Inc. Condenser, reflux, bulb, 29/42 outer top, 29/42 inner drip-tip bottom
SDP
Catalog No. 50972744
Encompass_Preferred
Bulb-style reflux condenser with 29/42 outer top joint and 29/42 inner drip-tip bottom joint.
Catalog No. 50-972-744 Supplier Ace Glass, Inc. Supplier No. 604206
Bulb-style reflux condenser with 29/42 outer top joint and 29/42 inner drip-tip bottom joint.