Learn More
Chemglass Life Sciences Condenser, Reflux, Double Cooling, 320 x 45mm, 24/40 Joint
SDP
Catalog No. 50143895
Encompass_Preferred
Condenser, Reflux, Double Cooling, 320 x 45mm, 24/40 Joint. High efficiency reflux condenser employs a double surface area with a cooling finger sealed into the secondary cooling jacket. Serrated hose connections for water connection extend through the outer bulb and have an O.D. of 10mm at the middle serration. The jacket length is 200mm.PLEASE NOTE: THIS CONDENSER DESIGN WORKS BEST WITH CLOSED LOOP CHILLERS.
Catalog No. 50-143-895 Supplier Chemglass Life Sciences Supplier No. CG1217A24
Condenser, Reflux, Double Cooling, 320 x 45mm, 24/40 Joint. High efficiency reflux condenser employs a double surface area with a cooling finger sealed into the secondary cooling jacket. Serrated hose connections for water connection extend through the outer bulb and have an O.D. of 10mm at the middle serration. The jacket length is 200mm.PLEASE NOTE: THIS CONDENSER DESIGN WORKS BEST WITH CLOSED LOOP CHILLERS.