Learn More
Ace Glass, Inc. Condenser, 460mm, 46in Soxhlet apparatus, 103/60 bottom, 22mm OD tube end top, size F hose connections for 7/16 or 1/2 inch ID tubing
SDP
Catalog No. 50973422
Encompass_Preferred
Catalog No. 50-973-422 Supplier Ace Glass, Inc. Supplier No. 681014